siRNA Details
Gene Symbol: CNTLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114395.2 | NP_001107867.1 | 5 | 840 | ||
NM_017738.3 | NP_060208.2 | 5 | 840 | |||
XM_005251492.1 | XP_005251549.1 | 5 | 840 | |||
XM_006716793.3 | XP_006716856.1 | 5 | 638 | |||
XM_011517941.2 | XP_011516243.1 | 5 | 840 | |||
XM_017014839.1 | XP_016870328.1 | 5 | 840 | |||
XM_017014840.1 | XP_016870329.1 | 5 | 840 | |||
XM_017014841.1 | XP_016870330.1 | 5 | 840 | |||
XM_017014842.1 | XP_016870331.1 | 5 | 840 | |||
XM_017014843.1 | XP_016870332.1 | 5 | 840 | |||
GenBank mRNA | AK098502.1 | - | 5 | 791 | ||
AK308837.1 | - | 5 | 795 | |||
BX647069.1 | - | 5 | 798 |
