siRNA Details
Gene Symbol: FAM120C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017848.5 | NP_060318.4 | 12 | 2554 | ||
XM_006724589.3 | XP_006724652.1 | 12 | 2550 | |||
GenBank mRNA | AY150025.1 | - | 12 | 2554 | ||
BC136413.1 | - | 12 | 2551 | 9020401 |
