siRNA Details
Gene Symbol: C1orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017891.4 | NP_060361.4 | 9, 10 | 941 | ||
XM_017001579.1 | XP_016857068.1 | 9 | 2044 | |||
XM_017001580.1 | XP_016857069.1 | 11, 12 | 2134 | |||
GenBank mRNA | AK000591.1 | - | 9, 10 | 676 | ||
AK054793.1 | - | 8, 9 | 1776 | |||
AK128271.1 | - | 3 | 2159 | |||
BC008788.2 | - | 8, 9 | 774 | 3627081 | ||
BE280321.1 | - | 3, 4 | 131 |
