siRNA Details
Gene Symbol: SRBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018079.4 | NP_060549.4 | 12 | 1680 | ||
XM_011532946.1 | XP_011531248.1 | 12 | 1681 | |||
XM_017004417.1 | XP_016859906.1 | 12 | 1681 | |||
XM_017004418.1 | XP_016859907.1 | 4 | 1282 | |||
GenBank mRNA | AK001241.1 | - | 6 | 577 | ||
AK056536.1 | - | 12 | 1650 | |||
AK301122.1 | - | 4 | 379 | |||
AK316222.1 | - | 4 | 360 | |||
BC032538.1 | - | 6 | 655 | 5528417 |
