siRNA Details
Gene Symbol: C5orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018356.2 | NP_060826.2 | 3 | 380 | ||
NR_134298.1 | 3 | 380 | ||||
XM_006714479.2 | XP_006714542.1 | 3 | 262 | |||
XM_006714480.3 | XP_006714543.1 | 3 | 270 | |||
XM_011514062.2 | XP_011512364.1 | 4 | 458 | |||
XM_017009607.1 | XP_016865096.1 | 4 | 462 | |||
XM_017009608.1 | XP_016865097.1 | 3 | 380 | |||
XM_017009609.1 | XP_016865098.1 | 3 | 262 | |||
XM_017009610.1 | XP_016865099.1 | 3 | 369 | |||
XM_017009611.1 | XP_016865100.1 | 3 | 369 | |||
XM_017009612.1 | XP_016865101.1 | 3 | 278 | |||
XM_017009613.1 | XP_016865102.1 | 3 | 560 | |||
XM_017009614.1 | XP_016865103.1 | 4 | 455 | |||
XM_017009615.1 | XP_016865104.1 | 4 | 363 | |||
XM_017009616.1 | XP_016865105.1 | 3 | 277 | |||
GenBank mRNA | AK002055.1 | - | 3 | 367 | ||
AK299156.1 | - | 3 | 357 | |||
BC021215.2 | - | 3 | 317 | 4302543 | ||
BC032845.1 | - | 3 | 363 | |||
DA783320.1 | - | 3 | 262 | |||
DC404356.1 | - | 3 | 380 |
