siRNA Details
Gene Symbol: SPATA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040428.3 | NP_001035518.1 | 6, 7 | 989 | ||
NM_018418.4 | NP_060888.2 | 7, 8 | 1085 | |||
XM_017021452.1 | XP_016876941.1 | 7, 8 | 1250 | |||
XM_017021454.1 | XP_016876943.1 | 8, 9 | 1118 | |||
XM_017021455.1 | XP_016876944.1 | 7, 8 | 1051 | |||
XM_017021456.1 | XP_016876945.1 | 8, 9 | 1064 | |||
XM_017021457.1 | XP_016876946.1 | 6, 7 | 1189 | |||
GenBank mRNA | AF144487.3 | - | 7, 8 | 1065 | ||
AK290631.1 | - | 6, 7 | 960 | |||
AL136604.1 | - | 6, 7 | 969 | |||
BC008656.1 | - | 2, 3 | 517 | |||
BC090875.1 | - | 7, 8 | 1372 | 30383306 | ||
CR533563.1 | - | 6, 7 | 814 | |||
CR936777.1 | - | 7, 8 | 1031 | |||
EU668353.1 | - | 7, 8 | 1061 |
