siRNA Details
Gene Symbol: AJAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042478.1 | NP_001035943.1 | 2 | 956 | ||
NM_018836.3 | NP_061324.1 | 2 | 956 | |||
XM_011541786.2 | XP_011540088.1 | 2 | 1226 | |||
XM_011541787.2 | XP_011540089.1 | 2 | 1112 | |||
GenBank mRNA | AF175409.1 | - | 2 | 1268 | ||
AL049672.1 | - | 1 | 41 | |||
AL049673.1 | - | 1 | 41 | |||
AY282806.1 | - | 2 | 956 |
