siRNA Details
Gene Symbol: PCDHB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018940.3 | NP_061763.1 | 1 | 902 | ||
GenBank mRNA | AF152500.1 | - | 1 | 687 | ||
AF217750.1 | - | 1 | 687 | |||
AK024641.1 | - | 1 | 861 | |||
AK222560.1 | - | 1 | 863 | |||
AK222678.1 | - | 1 | 847 | |||
AK315054.1 | - | 1 | 905 | |||
AL834254.1 | - | 1 | 249 | |||
BC098574.1 | - | 1 | 841 | 30925247 | ||
BC130331.1 | - | 1 | 769 | 40146362 | ||
BC130333.1 | - | 1 | 769 | 40146364 |
