siRNA Details
Gene Symbol: MMP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021801.4 | NP_068573.2 | 1 | 39 | ||
GenBank mRNA | AF230354.1 | - | 1 | 23 | ||
AF248646.1 | - | 1 | 27 | |||
AF291664.1 | - | 1 | 36 | |||
AJ251531.1 | - | 1 | 33 | |||
BC101541.1 | - | 1 | 39 | 8069047 | ||
BC101543.1 | - | 1 | 39 | 8069049 |
