siRNA Details
Gene Symbol: CD177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020406.3 | NP_065139.2 | 9 | 1356 | ||
GenBank mRNA | AB237911.1 | - | 9 | 1308 | ||
AF146747.1 | - | 9 | 1335 | |||
AJ290452.1 | - | 9 | 1327 | |||
AJ305326.1 | - | 9 | 1445 | |||
AK225226.1 | - | 9 | 1329 | |||
AK290965.1 | - | 9 | 1331 | |||
AY358932.1 | - | 9 | 1316 | |||
BC029167.1 | - | 9 | 1318 | 5182826 |
