siRNA Details
Gene Symbol: ZNF286A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130842.1 | NP_001124314.1 | 5 | 660 | ||
NM_001288642.1 | NP_001275571.1 | 5 | 587 | |||
NM_001288643.1 | NP_001275572.1 | 4 | 428 | |||
NM_001288644.1 | NP_001275573.1 | 3 | 313 | |||
NM_001288645.1 | NP_001275574.1 | 5 | 747 | |||
NM_001288646.1 | NP_001275575.1 | 5 | 660 | |||
NM_001288647.1 | NP_001275576.1 | 4 | 545 | |||
NM_001288648.1 | NP_001275577.1 | 4 | 428 | |||
NM_001288649.1 | NP_001275578.1 | 3 | 249 | |||
NM_020652.2 | NP_065703.1 | 5 | 747 | |||
GenBank mRNA | AF086305.1 | - | 3 | 180 | ||
AF217226.1 | - | 4 | 579 | |||
AK092259.1 | - | 5 | 585 | |||
AK315062.1 | - | 5 | 505 | |||
BC015730.1 | - | 5 | 585 | |||
BC025260.1 | - | 5 | 585 | |||
DA096356.1 | - | 4 | 251 | |||
DA748526.1 | - | 5 | 498 | |||
DA786711.1 | - | 3 | 87 | |||
DA796080.1 | - | 5 | 420 | |||
DC344287.1 | - | 4 | 314 | |||
DN997307.1 | - | 4 | 443 |
