siRNA Details
Gene Symbol: ZNF304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290318.1 | NP_001277247.1 | 4 | 1322 | ||
NM_001290319.1 | NP_001277248.1 | 4 | 1262 | |||
NM_020657.3 | NP_065708.2 | 3 | 1181 | |||
XM_011527145.2 | XP_011525447.1 | 5 | 1424 | |||
XM_017027024.1 | XP_016882513.1 | 4 | 1714 | |||
GenBank mRNA | AJ276316.1 | - | 4 | 1076 | ||
BC111531.1 | - | 4 | 1210 | 40012297 | ||
BC111532.1 | - | 4 | 1270 | 40012300 |
