siRNA Details
Gene Symbol: KIAA1324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267048.1 | NP_001253977.1 | 4 | 774 | ||
NM_001284352.1 | NP_001271281.1 | 3 | 653 | |||
NM_020775.4 | NP_065826.2 | 4 | 774 | |||
XM_011541825.1 | XP_011540127.1 | 4 | 634 | |||
XM_011541826.2 | XP_011540128.1 | 4 | 634 | |||
XM_011541827.1 | XP_011540129.1 | 4 | 634 | |||
XM_017001883.1 | XP_016857372.1 | 4 | 721 | |||
GenBank mRNA | AK057647.1 | - | 4 | 632 | ||
AK297221.1 | - | 3 | 447 | |||
AK304003.1 | - | 3 | 443 | |||
AY358366.1 | - | 5 | 690 | |||
BC125208.1 | - | 4 | 582 | 40123480 | ||
CA775143.1 | - | 4 | 600 | |||
CD360008.1 | - | 2 | 136 | |||
DA998586.1 | - | 2 | 142 |
