siRNA Details
Gene Symbol: KIAA1522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198972.1 | NP_001185901.1 | 3 | 402 | ||
NM_001198973.1 | NP_001185902.1 | 3 | 402 | |||
NM_020888.2 | NP_065939.2 | 3 | 487 | |||
XM_006710791.3 | XP_006710854.1 | 3 | 421 | |||
XM_017001916.1 | XP_016857405.1 | 1 | 28 | |||
XM_017001917.1 | XP_016857406.1 | 3 | 559 | |||
GenBank mRNA | AB040955.1 | - | 3 | 369 | ||
AK298965.1 | - | 3 | 402 |
