siRNA Details
Gene Symbol: PWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005049.2 | NP_005040.2 | 16 | 2091 | ||
XM_011529667.2 | XP_011527969.1 | 16 | 2156 | |||
GenBank mRNA | AB209438.1 | - | 5 | 3535 | ||
AK314184.1 | - | 16 | 2095 | |||
BC013309.2 | - | 16 | 2001 | 3528790 | ||
BC014988.1 | - | 16 | 2001 | 4906611 | ||
BX387451.2 | - | 4 | 401 | |||
U53346.1 | - | 16 | 2025 | |||
U56085.1 | - | 16 | 2036 | |||
X95263.1 | - | 16 | 2032 |
Gene Symbol: LOC102724159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723910.1 | XP_006723973.1 | 16 | 2128 |
