siRNA Details
Gene Symbol: ZNF350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021632.3 | NP_067645.3 | 5 | 694 | ||
XM_011527196.1 | XP_011525498.1 | 5 | 738 | |||
XM_017027094.1 | XP_016882583.1 | 6 | 850 | |||
XM_017027095.1 | XP_016882584.1 | 6 | 1011 | |||
XM_017027096.1 | XP_016882585.1 | 6 | 871 | |||
XM_017027097.1 | XP_016882586.1 | 6 | 884 | |||
XM_017027098.1 | XP_016882587.1 | 6 | 971 | |||
XM_017027099.1 | XP_016882588.1 | 7 | 994 | |||
XM_017027100.1 | XP_016882589.1 | 5 | 853 | |||
XM_017027101.1 | XP_016882590.1 | 6 | 837 | |||
XM_017027102.1 | XP_016882591.1 | 6 | 883 | |||
XM_017027103.1 | XP_016882592.1 | 6 | 971 | |||
XM_017027104.1 | XP_016882593.1 | 6 | 975 | |||
GenBank mRNA | AF295096.1 | - | 4 | 649 | ||
AF309561.1 | - | 5 | 683 | |||
AK023467.1 | - | 5 | 694 | |||
BC009921.2 | - | 4 | 634 | 3506774 |
