siRNA Details
Gene Symbol: RSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012425.3 | NP_036557.1 | 4 | 392 | ||
NM_152724.2 | NP_689937.2 | 3 | 280 | |||
XM_005252552.3 | XP_005252609.1 | 4 | 475 | |||
GenBank mRNA | AK225681.1 | - | 4 | 334 | ||
AK292911.1 | - | 3 | 205 | |||
AK312520.1 | - | 4 | 335 | |||
BC005993.1 | - | 4 | 362 | 4050837 | ||
BC008691.1 | - | 3 | 551 | 3872071 | ||
BC015644.2 | - | 4 | 352 | 4647836 | ||
BC107898.1 | - | 3 | 259 | |||
BG400752.1 | - | 3 | 281 | |||
BX647733.1 | - | 3 | 256 | |||
CR536521.1 | - | 3 | 249 | |||
CR541840.1 | - | 3 | 249 |
