siRNA Details
Gene Symbol: SECTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003004.2 | NP_002995.1 | 3 | 772 | ||
XM_005256392.3 | XP_005256449.1 | 3 | 556 | |||
XM_011523588.2 | XP_011521890.1 | 3 | 803 | |||
GenBank mRNA | AK223068.1 | - | 3 | 465 | ||
AK223104.1 | - | 3 | 496 | |||
AK290708.1 | - | 3 | 725 | |||
AK312980.1 | - | 3 | 492 | |||
BC017716.1 | - | 3 | 466 | 4413387 | ||
BI861812.1 | - | 1 | 193 | |||
U77643.1 | - | 3 | 492 |
