siRNA Details
Gene Symbol: ECSCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077693.3 | NP_001071161.1 | 2, 3 | 187 | ||
NM_001293739.1 | NP_001280668.1 | 2, 3 | 187 | |||
NR_121659.1 | 2, 3 | 187 | ||||
GenBank mRNA | BC146912.1 | - | 2, 3 | 159 | 9056840 | |
BC146913.1 | - | 2, 3 | 159 | 9056841 | ||
BF526332.1 | - | 207 | ||||
BI834795.1 | - | 2, 3 | 137 | |||
DQ462572.1 | - | 2, 3 | 181 | |||
EU025066.1 | - | 2, 3 | 187 |
