siRNA Details
Gene Symbol: FAM110C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077710.2 | NP_001071178.2 | 1 | 853 | ||
XM_011510372.2 | XP_011508674.1 | 1 | 855 | |||
XM_017004689.1 | XP_016860178.1 | 2 | 1047 | |||
XM_017004690.1 | XP_016860179.1 | 2 | 795 | |||
XM_017004691.1 | XP_016860180.1 | 1 | 855 | |||
XM_017004692.1 | XP_016860181.1 | 1 | 855 | |||
GenBank mRNA | AK302544.1 | - | 1 | 794 | ||
DQ431183.1 | - | 1 | 650 |
