siRNA Details
Gene Symbol: MROH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100878.1 | NP_001094348.1 | 1 | 188 | ||
XM_006716615.2 | XP_006716678.1 | 1 | 182 | |||
XM_011517214.1 | XP_011515516.1 | 1 | 182 | |||
XM_011517215.1 | XP_011515517.1 | 1 | 182 | |||
XM_011517216.2 | XP_011515518.1 | 1 | 182 | |||
XM_011517217.1 | XP_011515519.1 | 1 | 182 | |||
XM_011517218.1 | XP_011515520.1 | 1 | 182 | |||
XM_011517219.1 | XP_011515521.1 | 1 | 182 | |||
XM_011517220.1 | XP_011515522.1 | 1 | 182 | |||
XM_011517221.2 | XP_011515523.1 | 1 | 182 | |||
XM_011517222.1 | XP_011515524.1 | 1 | 182 | |||
XM_011517223.1 | XP_011515525.1 | 1 | 182 | |||
GenBank mRNA | BM148864.1 | - | 1 | 202 | ||
BX341537.2 | - | 1 | 187 |
