siRNA Details
Gene Symbol: MIR205HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104548.1 | NP_001098018.1 | 2 | 474 | ||
XM_017002058.1 | XP_016857547.1 | 1 | 569 | |||
GenBank mRNA | AK091113.1 | - | 1 | 569 | ||
BG674039.1 | - | 2 | 458 | |||
BM980083.1 | - | 3 | 429 | |||
CV575609.1 | - | 2 | 461 | |||
DA461662.1 | - | 2 | 473 |
