siRNA Details
Gene Symbol: MIR205HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104548.1 | NP_001098018.1 | 4 | 576 | ||
XM_017002058.1 | XP_016857547.1 | 3 | 671 | |||
GenBank mRNA | AK091113.1 | - | 2 | 3037 | ||
AW275825.1 | - | 1 | 308 | |||
BG674039.1 | - | 5 | 668 | |||
BM980083.1 | - | 1 | 327 |
