siRNA Details
Gene Symbol: TMEM253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146683.1 | NP_001140155.1 | 3 | 493 | ||
XM_006720233.3 | XP_006720296.1 | 2 | 1068 | |||
XM_011537078.2 | XP_011535380.1 | 3 | 370 | |||
XM_011537079.1 | XP_011535381.1 | 2 | 212 | |||
XM_011537081.2 | XP_011535383.1 | 2 | 1070 | |||
XM_011537082.2 | XP_011535384.1 | 3 | 488 | |||
XM_011537083.2 | XP_011535385.1 | 2 | 1071 | |||
XM_011537084.2 | XP_011535386.1 | 2 | 1079 | |||
XM_011537085.2 | XP_011535387.1 | 2 | 1074 |
