siRNA Details
Gene Symbol: IKZF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022465.3 | NP_071910.3 | 2 | 729 | ||
XM_005269086.3 | XP_005269143.1 | 4 | 271 | |||
XM_011538664.1 | XP_011536966.1 | 4 | 274 | |||
XM_011538666.2 | XP_011536968.1 | 3 | 122 | |||
XM_017019805.1 | XP_016875294.1 | 6 | 519 | |||
XM_017019806.1 | XP_016875295.1 | 6 | 762 | |||
XM_017019807.1 | XP_016875296.1 | 6 | 759 | |||
XM_017019808.1 | XP_016875297.1 | 5 | 444 | |||
XM_017019809.1 | XP_016875298.1 | 6 | 516 | |||
XM_017019810.1 | XP_016875299.1 | 6 | 614 | |||
XM_017019811.1 | XP_016875300.1 | 5 | 441 | |||
XM_017019812.1 | XP_016875301.1 | 4 | 517 | |||
XM_017019813.1 | XP_016875302.1 | 4 | 514 | |||
GenBank mRNA | BX647761.1 | - | 3 | 299 | ||
DA797441.1 | - | 2 | 293 |
