siRNA Details
Gene Symbol: LOC643802 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207030.1 | NP_001193959.1 | 1 | 178 | ||
XM_017023559.1 | XP_016879048.1 | 1 | 261 | |||
XM_017023560.1 | XP_016879049.1 | 1 | 261 | |||
GenBank mRNA | AV757420.1 | - | 1 | 248 | ||
BX537874.1 | - | 1 | 19 |
