siRNA Details
Gene Symbol: SMIM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163438.1 | NP_001156910.1 | 1 | 187 | ||
GenBank mRNA | BC008642.1 | - | 1 | 187 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SMIM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163438.1 | NP_001156910.1 | 1 | 187 | ||
GenBank mRNA | BC008642.1 | - | 1 | 187 |