siRNA Details
Gene Symbol: C19orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277378.1 | NP_001264307.1 | 5, 6 | 1060 | ||
XM_011528184.2 | XP_011526486.1 | 5, 6 | 1415 | |||
GenBank mRNA | DN831393.1 | - | 3, 4 | 414 | ||
HY212999.1 | - | 1, 2 | 384 |
