siRNA Details
Gene Symbol: LOC646588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012912.1 | XP_016868401.1 | 3 | 346 | ||
XM_017012913.1 | XP_016868402.1 | 3 | 189 | |||
XM_017012914.1 | XP_016868403.1 | 3 | 189 | |||
XM_017012915.1 | XP_016868404.1 | 3 | 189 | |||
XM_017012916.1 | XP_016868405.1 | 3 | 189 | |||
XM_017012917.1 | XP_016868406.1 | 3 | 531 | |||
XM_017012918.1 | XP_016868407.1 | 3 | 189 | |||
GenBank mRNA | AK057379.1 | - | 3 | 398 |
