siRNA Details
Gene Symbol: SERPINE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101320.1 | NP_001094790.1 | 1 | 253 | ||
XM_017020707.1 | XP_016876196.1 | 1 | 6309 | |||
GenBank mRNA | AM402969.1 | - | 1 | 196 | ||
AX772926.1 | - | 1 | 253 |
