siRNA Details
Gene Symbol: GMNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146686.2 | NP_001140158.1 | 5 | 958 | ||
GenBank mRNA | BC031680.1 | - | 1 | 703 | ||
CR749823.1 | - | 2 | 791 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GMNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146686.2 | NP_001140158.1 | 5 | 958 | ||
GenBank mRNA | BC031680.1 | - | 1 | 703 | ||
CR749823.1 | - | 2 | 791 |