siRNA Details
Gene Symbol: SMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258423.1 | NP_001245352.1 | 5, 6 | 837 | ||
NM_004595.4 | NP_004586.2 | 7, 8 | 996 | |||
XM_005274582.2 | XP_005274639.1 | 7, 8 | 1062 | |||
XM_011545568.2 | XP_011543870.1 | 7, 8 | 856 | |||
XM_017029753.1 | XP_016885242.1 | 7, 8 | 836 | |||
XM_017029754.1 | XP_016885243.1 | 7, 8 | 803 | |||
XM_017029755.1 | XP_016885244.1 | 7, 8 | 894 | |||
GenBank mRNA | AD001528.1 | - | 7, 8 | 845 | ||
AK293455.1 | - | 6, 7 | 740 | |||
AK313834.1 | - | 7, 8 | 828 | |||
BC009898.2 | - | 7, 8 | 819 | 3506022 | ||
BC085621.1 | - | 7, 8 | 788 | 5540463 | ||
Z49099.1 | - | 7, 8 | 772 |
