siRNA Details
Gene Symbol: TAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135218.1 | NP_001128690.1 | 2 | 552 | ||
NM_005644.3 | NP_005635.1 | 2 | 215 | |||
XM_017002186.1 | XP_016857675.1 | 2 | 617 | |||
GenBank mRNA | AK310097.1 | - | 2 | 209 | ||
BC011986.1 | - | 2 | 465 | 3882343 | ||
BE787487.1 | - | 2 | 444 | |||
BT007031.1 | - | 1 | 30 | |||
BX095156.1 | - | 2 | 215 | |||
CX865268.1 | - | 2 | 474 | |||
D50544.1 | - | 1 | 116 | |||
U57693.1 | - | 2 | 155 | |||
X84002.1 | - | 2 | 196 |
