siRNA Details
Gene Symbol: LINC01193 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040094.1 | 4, 5 | 711 | |||
GenBank mRNA | BC047459.1 | - | 4, 5 | 711 |
Gene Symbol: LOC101927079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110480.1 | 2, 3 | 224 | |||
GenBank mRNA | AK058056.1 | - | 2, 3 | 224 | ||
DB049942.1 | - | 2, 3 | 224 |
Gene Symbol: LOC102723532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546205.1 | XP_011544507.1 | 2, 3 | 224 | ||
XM_011546206.1 | XP_011544508.1 | 2, 3 | 224 |
Gene Symbol: LOC727924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015416.1 | 2, 3 | 223 | |||
GenBank mRNA | BC048276.1 | - | 2, 3 | 223 |
