siRNA Details
Gene Symbol: LOC100505841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195535.1 | NP_001182464.1 | 2 | 113 | ||
XM_011543086.2 | XP_011541388.1 | 5 | 432 | |||
GenBank mRNA | AI188208.1 | - | 2 | 328 | ||
BU567915.1 | - | 2 | 116 |
