siRNA Details
Gene Symbol: DNLZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080849.2 | NP_001074318.1 | 2 | 448 | ||
NR_073565.1 | 2 | 452 | ||||
GenBank mRNA | AI435052.1 | - | 2 | 293 | ||
AI696701.1 | - | 2 | 299 | |||
BC146915.1 | - | 2 | 448 | 9056843 | ||
BC146924.1 | - | 2 | 448 | 9056852 |
