siRNA Details
Gene Symbol: SKP1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036619.1 | 1 | 755 | |||
GenBank mRNA | AK310444.1 | - | 1 | 755 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SKP1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036619.1 | 1 | 755 | |||
GenBank mRNA | AK310444.1 | - | 1 | 755 |