siRNA Details
Gene Symbol: CCDC61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267723.1 | NP_001254652.1 | 2, 3 | 194 | ||
XM_005259191.3 | XP_005259248.1 | 2, 3 | 2299 | |||
XM_005259192.4 | XP_005259249.1 | 2, 3 | 940 | |||
XM_011527257.2 | XP_011525559.1 | 2, 3 | 223 | |||
GenBank mRNA | DR007040.1 | - | 2, 3 | 214 | ||
GQ375050.1 | - | 2, 3 | 167 |
