siRNA Details
Gene Symbol: LOC102724467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120309.1 | 1 | 73 | |||
GenBank mRNA | DB094384.1 | - | 1 | 75 | ||
DB100335.1 | - | 1 | 299 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102724467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120309.1 | 1 | 73 | |||
GenBank mRNA | DB094384.1 | - | 1 | 75 | ||
DB100335.1 | - | 1 | 299 |