siRNA Details
Gene Symbol: LOC102724279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302493.1 | NP_001289422.1 | 3 | 209 | ||
GenBank mRNA | AI436266.1 | - | 1 | 241 | ||
BX095604.1 | - | 3 | 209 | |||
GD139371.1 | - | 3 | 184 |
