siRNA Details
Gene Symbol: LOC100996630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135588.1 | 1 | 73 | |||
GenBank mRNA | AI018412.1 | - | 2 | 306 | ||
BX417179.2 | - | 1 | 73 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100996630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135588.1 | 1 | 73 | |||
GenBank mRNA | AI018412.1 | - | 2 | 306 | ||
BX417179.2 | - | 1 | 73 |