siRNA Details
Gene Symbol: ZNF726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244038.1 | NP_001230967.1 | 3 | 308 | ||
GenBank mRNA | AB593087.1 | - | 3 | 271 | ||
BC046415.1 | - | 3 | 280 | 4821872 | ||
BE783649.1 | - | 3 | 280 | |||
BX475164.1 | - | 3 | 316 | |||
CN359557.1 | - | 3 | 308 |
Gene Symbol: LOC100505851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125686.1 | - | 3 | 612 |
