siRNA Details
Gene Symbol: FAM166B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099951.3 | NP_001093421.1 | 3 | 458 | ||
NM_001164310.2 | NP_001157782.1 | 3 | 458 | |||
NM_001287238.1 | NP_001274167.1 | 3 | 458 | |||
NM_001287239.1 | NP_001274168.1 | 3 | 458 | |||
XM_011518028.2 | XP_011516330.1 | 3 | 597 | |||
GenBank mRNA | BC129999.1 | - | 3 | 458 | 40127800 | |
BC146928.1 | - | 3 | 458 | 9056856 | ||
BC146933.1 | - | 3 | 458 | 9056861 | ||
BC171840.1 | - | 3 | 458 | 9054494 | ||
BC171851.1 | - | 3 | 458 | 9054505 | ||
CB241755.1 | - | 4 | 631 |
