siRNA Details
Gene Symbol: LOC100507073 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110156.1 | 5 | 354 | |||
GenBank mRNA | CN360491.1 | - | 5 | 354 | ||
DA610966.1 | - | 2 | 151 | |||
DA614164.1 | - | 2 | 159 | |||
DA614343.1 | - | 2 | 162 | |||
DA616181.1 | - | 2 | 159 | |||
DA618279.1 | - | 2 | 144 |
