siRNA Details
Gene Symbol: LOC100507073 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110156.1 | 5, 6 | 382 | |||
GenBank mRNA | CN360491.1 | - | 5, 6 | 382 | ||
DA610966.1 | - | 2, 3 | 179 | |||
DA614343.1 | - | 2, 3 | 190 | |||
DA616181.1 | - | 2, 3 | 187 |
