siRNA Details
Gene Symbol: UPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006760.3 | NP_006751.1 | 2, 3 | 264 | ||
GenBank mRNA | AB009454.1 | - | 2, 3 | 151 | ||
AY455312.1 | - | 2, 3 | 211 | |||
BC113900.1 | - | 2, 3 | 242 | 40029623 | ||
CD556028.1 | - | 2, 3 | 279 | |||
Y13645.1 | - | 2, 3 | 249 |
