siRNA Details
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 1174 | ||
NR_036599.1 | 5 | 1292 | ||||
NR_036600.1 | 3 | 1047 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 1149 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 1204 | |||
GenBank mRNA | AK131368.1 | - | 2 | 1772 | ||
AK131574.1 | - | 1 | 559 | |||
AK293074.1 | - | 3 | 1047 | |||
AK304704.1 | - | 4 | 1138 | |||
BC140720.1 | - | 3 | 1075 | 9021588 | ||
BX640767.1 | - | 1 | 201 | |||
CR933598.1 | - | 4 | 1137 |
