siRNA Details
Gene Symbol: MZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003422.2 | NP_003413.2 | 6 | 1062 | ||
NM_198055.1 | NP_932172.1 | 6 | 1380 | |||
XM_005259204.3 | XP_005259261.1 | 7 | 1735 | |||
XM_011527264.2 | XP_011525566.1 | 6 | 1464 | |||
XM_017027206.1 | XP_016882695.1 | 6 | 1583 | |||
GenBank mRNA | AF055077.1 | - | 6 | 1062 | ||
AF055078.1 | - | 6 | 1380 | |||
BC007777.2 | - | 6 | 1038 | 4301723 | ||
M58297.1 | - | 1 | 1162 |
