siRNA Details
Gene Symbol: ZNF148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021964.2 | NP_068799.2 | 9 | 2358 | ||
GenBank mRNA | AF039019.1 | - | 8 | 2083 | ||
AF432210.1 | - | 9 | 2174 | |||
AJ236885.1 | - | 9 | 2263 | |||
AK314203.1 | - | 9 | 2293 | |||
AL833616.1 | - | 9 | 2196 | |||
BC050260.1 | - | 10 | 2434 | 4423572 | ||
DQ090088.1 | - | 6 | 1868 | |||
L04282.1 | - | 9 | 2256 |
