siRNA Details
Gene Symbol: FNDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319152.1 | NP_001306081.1 | 2 | 431 | ||
NM_001319153.1 | NP_001306082.1 | 2 | 433 | |||
NM_024059.3 | NP_076964.1 | 2 | 449 | |||
XM_011529041.2 | XP_011527343.1 | 2 | 986 | |||
XM_017028060.1 | XP_016883549.1 | 2 | 585 | |||
GenBank mRNA | AK125168.1 | - | 1 | 2306 | ||
BC000912.2 | - | 2 | 449 | 3048411 | ||
BG772889.1 | - | 2 | 445 | |||
BI828873.1 | - | 2 | 426 | |||
BM554007.1 | - | 2 | 410 | |||
BQ109512.1 | - | 2 | 432 |
